1-Bromo-4-ethoxy-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

20428-25G 20428-5G
76884-584EA 346.92 USD
76884-584 76884-586
1-Bromo-4-ethoxy-2-fluorobenzene ≥97%
1-Bromo-4-ethoxy-2-fluorobenzene
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD18447607
CAS Number: 107713-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR