3-Bromo-6-chloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-6-chloro-2-fluorophenyl)methanol

20407-1G 20407-25G 20407-5G 20407-500MG
76884-574EA 106.09 USD
76884-574 76884-576 76884-578 77645-162
3-Bromo-6-chloro-2-fluorobenzyl alcohol ≥97%
3-Bromo-6-chloro-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447606
CAS Number: 886615-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR