1-Bromo-4,5-difluoro-2-ethoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

20383-25G 20383-5G
76884-566EA 559.1 USD
76884-566 76884-568
1-Bromo-4,5-difluoro-2-ethoxybenzene ≥97% research grade
1-Bromo-4,5-difluoro-2-ethoxybenzene
Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD18447604
CAS Number: 1266253-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR