1,2-Difluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20375-25G 20375-5G
76884-556EA 346.92 USD
76884-556 76884-558
1,2-Difluoro-3-isopropoxybenzene ≥97%
1,2-Difluoro-3-isopropoxybenzene
Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519048
CAS Number: 1174005-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR