1-Bromo-4-fluoro-2-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

20332-25G 20332-5G
76884-530EA 537.88 USD
76884-530 76884-532
1-Bromo-4-fluoro-2-isopropoxybenzene ≥95%
1-Bromo-4-fluoro-2-isopropoxybenzene
Formula: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD10699697
CAS Number: 610797-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR