1-Bromo-4-butoxy-2-fluorobenzene 97%

Supplier: AOBChem USA

Warning

20296-25G 20296-5G
76884-514EA 169.75 USD
76884-514 77634-316
1-Bromo-4-butoxy-2-fluorobenzene 97%
1-Bromo-4-butoxy-2-fluorobenzene
Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011995
CAS Number: 155854-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR