4-Bromo-3-methylthioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-methyl-4-(methylthio)benzene

20284-1G 20284-25G 20284-500MG 20284-5G
76884-500EA 124.13 USD
76884-500 76884-502 76884-504 76884-506
4-Bromo-3-methylthioanisole ≥97%
4-Bromo-3-methylthioanisole

Precaución: for Research Use Only

Formula: C₈H₉BrS
Storage Temperature: Refrigerator
MDL Number: MFCD09801036
CAS Number: 90532-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR