1-Bromo-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromobenzyl methyl ether

20232-25G 20232-5G
76884-476EA 219.61 USD
76884-476 76884-478
1-Bromo-2-(methoxymethyl)benzene ≥97%
1-Bromo-2-(methoxymethyl)benzene
Formula: C₈H₉BrO
MW: 201.0605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03790887
CAS Number: 52711-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR