1-Bromo-4-chloro-3-methylthio-2-fluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
20169-1G
20169-25G
20169-5G
76884-466EA
62.6
USD
76884-466
76884-468
76884-470
1-Bromo-4-chloro-3-methylthio-2-fluorobenzene ≥97%
1-Bromo-4-chloro-3-methylthio-2-fluorobenzene
Formula:
C₇H₅BrClFS Storage Temperature: Ambient |
MDL Number:
MFCD28402075 CAS Number: 943830-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...