1-Bromo-2,4-dichloro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20163-1G 20163-25G 20163-500MG 20163-5G
76884-452EA 171.87 USD
76884-452 76884-454 76884-456 76884-458
1-Bromo-2,4-dichloro-3-methoxybenzene ≥97%
1-Bromo-2,4-dichloro-3-methoxybenzene
Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447601
CAS Number: 174913-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR