1-Bromo-2-chloro-3,4-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

19978-1G 19978-25G 19978-500MG 19978-5G
76884-414EA 290.69 USD
76884-414 76884-416 76884-418 76884-420
1-Bromo-2-chloro-3,4-difluorobenzene ≥95%
1-Bromo-2-chloro-3,4-difluorobenzene
Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845914
CAS Number: 1160574-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR