1-Bromo-3,4-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

19976-1G 19976-25G 19976-500MG 19976-5G
76884-400EA 381.93 USD
76884-400 76884-402 76884-404 76884-406
1-Bromo-3,4-difluoro-2-iodobenzene ≥97%
1-Bromo-3,4-difluoro-2-iodobenzene
Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922588
CAS Number: 1208077-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR