1-Bromo-4-(tert-butyl)-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-(1,1-dimethylethyl)-2-iodo-benzene

19948-1G 19948-25G 19948-5G
76884-390EA 62.6 USD
76884-390 76884-392 76884-394
1-Bromo-4-(tert-butyl)-2-iodobenzene ≥97%
1-Bromo-4-(tert-butyl)-2-iodobenzene
Formula: C₁₀H₁₂BrI
MW: 339.0108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28400709
CAS Number: 916747-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR