1-Bromo-3-chloro-2-ethoxy-benzene ≥97%

Supplier: AOBChem USA

Warning

19935-1G 19935-25G 19935-5G 19935-500MG
76884-376EA 112.46 USD
76884-376 76884-378 76884-380 77645-126
1-Bromo-3-chloro-2-ethoxy-benzene ≥97%
1-Bromo-3-chloro-2-ethoxy-benzene
Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31382548
CAS Number: 1010390-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR