1-(Benzyloxy)-3-bromo-5-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

19909-1G 19909-25G 19909-5G
76884-366EA 66.84 USD
76884-366 76884-368 76884-370
1-(Benzyloxy)-3-bromo-5-chlorobenzene ≥97%
1-(Benzyloxy)-3-bromo-5-chlorobenzene
Formula: C₁₃H₁₀BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD16660296
CAS Number: 1187966-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR