2-Bromo-2,6-dichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

19905-1G 19905-25G 19905-5G 19905-500MG
76884-352EA 96.55 USD
76884-352 76884-354 76884-356 77649-190
2-Bromo-2,6-dichlorotoluene ≥97%
2-Bromo-2,6-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
Storage Temperature: Refrigerator
MDL Number: MFCD09801039
CAS Number: 115615-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR