1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Propylpyrazole-4-boronic acid Pinacol ester, 2-(1-Propyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Propyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

19831-25G 19831-5G
76884-342EA 304.48 USD
76884-342 76884-344
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97% research grade
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₂H₂₁BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD05663871
CAS Number: 827614-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR