1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Phenylpyrazole-4-boronic acid, pinacol ester, 1-Phenyl-1H-pyrazole-4-boronic acid pinacol ester

19616-1G 19616-25G 19616-500MG 19616-5G
76884-322EA 142.17 USD
76884-322 76884-324 76884-326 76884-328
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥95% research grade
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₅H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12400941
CAS Number: 1002334-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR