2,4-Dibromo-6-isopropylaniline ≥95%

Supplier: AOBChem USA

Warning

19498-25G 19498-5G
76884-302EA 310.85 USD
76884-302 76884-304
2,4-Dibromo-6-isopropylaniline ≥95%
2,4-Dibromo-6-isopropylaniline
Formula: C₉H₁₁Br₂N
MW: 292.9983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845854
CAS Number: 81090-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR