1,3-Dibromo-5-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

19419-25G 19419-5G
76884-274EA 187.78 USD
76884-274 76884-276
1,3-Dibromo-5-isopropylbenzene ≥97%
1,3-Dibromo-5-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂
MW: 277.9837 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657957
CAS Number: 62655-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR