2-(Benzyloxy)-4-methoxyphenylboronic acid 95%

Supplier: AOBChem USA

Warning

19363-1G 19363-500MG 19363-5G 19363-25G
76884-262EA 468.92 USD
76884-262 76884-264 76884-266 77635-518
2-(Benzyloxy)-4-methoxyphenylboronic acid 95%
2-(Benzyloxy)-4-methoxyphenylboronic acid
Formula: C₁₄H₁₅BO₄
MW: 258.077 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696653
CAS Number: 183474-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR