1-Bromo-3-fluoro-5-(propan-2-yloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

19218-1G 19218-25G 19218-500MG 19218-5G
76884-230EA 142.17 USD
76884-230 76884-232 76884-234 76884-236
1-Bromo-3-fluoro-5-(propan-2-yloxy)benzene ≥97%
1-Bromo-3-fluoro-5-(propan-2-yloxy)benzene
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917875
CAS Number: 1309933-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR