1-Ethylpyrazole-5-boronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Ethylpyrazole-5-boronic Acid Pinacol Ester, 1-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

19128-25G 19128-5G
76884-224EA 332.07 USD
76884-224 76884-226
1-Ethylpyrazole-5-boronic acid, pinacol ester ≥97%
1-Ethylpyrazole-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD16659009
CAS Number: 1007110-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR