(5-Ethyl-2-methoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

19113-1G 19113-25G 19113-500MG 19113-5G
76884-208EA 376.62 USD
76884-208 76884-210 76884-212 76884-214
(5-Ethyl-2-methoxyphenyl)boronic acid ≥97% research grade
(5-Ethyl-2-methoxyphenyl)boronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
CAS Number: 847345-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR