2,3-Difluoropyridine-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,3-Difluoropyridin-4-yl)boronic acid

19017-25G 19017-5G
76884-182EA 595.17 USD
76884-182 76884-184
2,3-Difluoropyridine-4-boronic acid ≥97%
2,3-Difluoropyridine-4-boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.899 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12022339
CAS Number: 1263374-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR