Benzo[b]thiophene-3-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzo[b]thiophen-3-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(Benzo[b]thiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

18998-25G 18998-5G
76884-172EA 233.4 USD
76884-172 76884-174
Benzo[b]thiophene-3-boronic acid pinacol ester ≥97% research grade
Benzo[b]thiophene-3-boronic acid pinacol ester
Formula: C₁₄H₁₇BO₂S
MW: 260.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110552
CAS Number: 171364-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR