2-(Benzyloxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

18986-1G 18986-25G 18986-5G 18986-500MG
76884-162EA 106.09 USD
76884-162 76884-164 76884-166 77645-066
2-(Benzyloxy)pyridine ≥97%
2-(Benzyloxy)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701434
CAS Number: 40864-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR