1-(4-Bromophenyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

18950-25G 18950-5G
76884-152EA 466.8 USD
76884-152 76884-154
1-(4-Bromophenyl)piperidine ≥97%
1-(4-Bromophenyl)piperidine
Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112533
CAS Number: 22148-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR