1-Methyl-1H-indazole-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methyl-1H-indazol-5-yl-5-boronic acid, (1-methyl-1H-indazol-5-yl)boronic acid

18944-25G 18944-5G
76884-142EA 346.92 USD
76884-142 76884-144
1-Methyl-1H-indazole-5-boronic acid ≥97%
1-Methyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09870052
CAS Number: 590418-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR