2-Acetyl-4-bromopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Bromo-pyridin-2-yl)-ethanone, 1-(4-Bromopyridin-2-yl)ethanone

18627-1G 18627-25G 18627-500MG 18627-5G
76884-082EA 150.65 USD
76884-082 76884-084 76884-086 76884-088
2-Acetyl-4-bromopyridine ≥97%
2-Acetyl-4-bromopyridine
Formula: C₇H₆BrNO
MW: 200.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189212
CAS Number: 1060805-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR