1-Bromo-3-chloro-4-(methylthio)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

18504-1G 18504-25G 18504-5G
76884-042EA 73.21 USD
76884-042 76884-044 76884-046
1-Bromo-3-chloro-4-(methylthio)benzene ≥97% research grade
1-Bromo-3-chloro-4-(methylthio)benzene
Formula: C₇H₆BrClS
MW: 237.5445 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543648
CAS Number: 101084-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR