2,4,6-Tribromobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
18457-25G
18457-5G
76884-030EA
456.19
USD
76884-030
76884-032
2,4,6-Tribromobenzoic acid ≥97%
2,4,6-Tribromobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃Br₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00059882 CAS Number: 633-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...