(5-Bromo-4-methyl-pyridin-3-yl)-methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
18414-1G
18414-25G
18414-500MG
18414-5G
76884-014EA
231.28
USD
76884-014
76884-016
76884-018
76884-020
(5-Bromo-4-methyl-pyridin-3-yl)-methanol ≥97%
(5-Bromo-4-methyl-pyridin-3-yl)-methanol
Formula:
C₇H₈BrNO MW: 202.049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13659373 CAS Number: 351458-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...