(2,3-Difluoro-4-isopropoxyphenyl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,3-Difluoro-4-isopropoxyphenylboronic acid
18393-1G
18393-25G
18393-5G
18393-500MG
76884-000EA
96.55
USD
76884-000
76884-002
76884-004
77649-054
(2,3-Difluoro-4-isopropoxyphenyl)boronic acid ≥97%
(2,3-Difluoro-4-isopropoxyphenyl)boronic acid
Formula:
C₉H₁₁BF₂O₃ MW: 215.99 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19237183 CAS Number: 1154762-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...