1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1,3,5-Trimethylpyrazole-4-boronic acid Pinacol ester, 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1,3,5-Trimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

18316-1G 18316-25G 18316-5G
76883-972EA 105.03 USD
76883-972 76883-974 76883-976
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₂₁BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06659062
CAS Number: 844891-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR