5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1H-Pyrazolo[3,4-b]pyridine-5-boronic acid, pinacol ester, 1H-Pyrazolo[3,4-b]pyridine-5-boronic acid pinacol ester

18303-1G 18303-25G 18303-5G
76883-958EA 109.28 USD
76883-958 76883-960 76883-962
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine ≥97% research grade
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine
Formula: C₁₂H₁₆BN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09832892
CAS Number: 1093819-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR