2-(3-Bromo-4-methylphenyl)-1,3-dioxolane 95%

Supplier: AOBChem USA

Warning

18162-1G 18162-25G 18162-5G 18162-500MG
76883-934EA 87 USD
76883-934 76883-936 76883-938 77649-036
2-(3-Bromo-4-methylphenyl)-1,3-dioxolane 95%
2-(3-Bromo-4-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27942538
CAS Number: 124717-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR