2,4,6-Trifluoro-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18132-1G 18132-25G 18132-500MG 18132-5G
76883-916EA 350.1 USD
76883-916 76883-918 76883-920 76883-922
2,4,6-Trifluoro-3-hydroxyphenylboronic acid ≥97%
2,4,6-Trifluoro-3-hydroxyphenylboronic acid
Formula: C₆H₄BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11053885
CAS Number: 1072951-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR