2,3-Difluoro-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

18089-25G 18089-5G
76883-900EA 622.75 USD
76883-900 76883-902
2,3-Difluoro-4-methylphenylboronic acid pinacol ester ≥97%
2,3-Difluoro-4-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BF₂O₂
MW: 254.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23379473
CAS Number: 1116681-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR