1-Bromo-2-fluoro-4,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

17968-1G 17968-25G 17968-500MG 17968-5G
76883-874EA 350.1 USD
76883-874 76883-876 76883-878 76883-880
1-Bromo-2-fluoro-4,5-dimethylbenzene ≥95%
1-Bromo-2-fluoro-4,5-dimethylbenzene
Formula: C₈H₈BrF
Storage Temperature: Refrigerator
MDL Number: MFCD08695022
CAS Number: 5100-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR