1H-Indazole-7-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

17858-1G 17858-25G 17858-5G
76883-846EA 92.3 USD
76883-846 76883-848 76883-850
1H-Indazole-7-boronic acid pinacol ester ≥97%
1H-Indazole-7-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08059264
CAS Number: 915411-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR