2,3-Difluoro-N-methoxy-N-methylbenzamide 97%

Supplier: AOBChem USA

Warning

17851-25G 17851-5G 17851-1G
76883-836EA 595.17 USD
76883-836 76883-838 77640-982
2,3-Difluoro-N-methoxy-N-methylbenzamide 97%
2,3-Difluoro-N-methoxy-N-methylbenzamide
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD11856049
CAS Number: 680610-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR