2-(2,3-Difluorophenyl)-2-methyl-[1,3]dioxolane ≥97%
Supplier: AOBChem USA
|
Warning
|
17849-25G
17849-5G
76883-830EA
346.92
USD
76883-830
76883-832
2-(2,3-Difluorophenyl)-2-methyl-[1,3]dioxolane ≥97%
2-(2,3-Difluorophenyl)-2-methyl-[1,3]dioxolane
Formula:
C₁₀H₁₀F₂O₂ MW: 200.182 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD25968416 CAS Number: 885590-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...