3-Methoxy-5-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid

17703-1G 17703-25G 17703-500MG 17703-5G
76883-818EA 158.08 USD
76883-818 76883-820 76883-822 76883-824
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid ≥97%
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₄
MW: 235.9529 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026733
CAS Number: 1256345-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR