1-Bromo-2-butoxy-5-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

17596-25G 17596-5G
76883-796EA 466.8 USD
76883-796 76883-798
1-Bromo-2-butoxy-5-fluorobenzene ≥95%
1-Bromo-2-butoxy-5-fluorobenzene
Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11127643
CAS Number: 1019369-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR