2,3-Dimethyl-5-bromofluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1-fluoro-2,3-dimethylbenzene

17586-1G 17586-500MG 17586-5G
76883-782EA 653.52 USD
76883-782 76883-784 76883-786
2,3-Dimethyl-5-bromofluorobenzene ≥97%
2,3-Dimethyl-5-bromofluorobenzene
Formula: C₈H₈BrF
MW: 203.052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02181182
CAS Number: 194805-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR