1-Bromo-2-(propan-2-yloxy)naphthalene ≥95%

Supplier: AOBChem USA

Warning

17528-1G 17528-25G 17528-500MG 17528-5G
76883-768EA 194.15 USD
76883-768 76883-770 76883-772 76883-774
1-Bromo-2-(propan-2-yloxy)naphthalene ≥95%
1-Bromo-2-(propan-2-yloxy)naphthalene
Formula: C₁₃H₁₃BrO
Storage Temperature: Refrigerator
MDL Number: MFCD12818581
CAS Number: 791088-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR