2-(4-Bromo-2-fluorophenoxy)propane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-1-isopropoxybenzene

17438-25G 17438-5G
76883-718EA 289.63 USD
76883-718 76883-720
2-(4-Bromo-2-fluorophenoxy)propane ≥97%
2-(4-Bromo-2-fluorophenoxy)propane
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070760
CAS Number: 202865-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR