1-(3-(benzyloxy)-4-fluorophenyl)ethanone ≥97% research grade

Supplier: AOBChem USA

Warning

17346-1G 17346-25G 17346-500MG 17346-5G
76883-702EA 167.63 USD
76883-702 76883-704 76883-706 76883-708
1-(3-(benzyloxy)-4-fluorophenyl)ethanone ≥97% research grade
1-(3-(benzyloxy)-4-fluorophenyl)ethanone
Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Storage Temperature: Refrigerator
CAS Number: 2167331-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR