2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17306-1G 17306-25G 17306-500MG 17306-5G
76883-680EA 379.81 USD
76883-680 76883-682 76883-684 76883-686
2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid ≥97%
2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BF₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11504848
CAS Number: 1072951-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR